Assay Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037937.3 | NP_001033026.1 | 9 | 6871 | 96 | |
| NM_145470.3 | NP_663445.2 | 9 | 7049 | 96 | ||
| GenBank mRNA | AK014624.1 | - | 1 | 651 | 96 | |
| AK171902.1 | - | 2815 | 96 | |||
| AK172487.1 | - | 1 | 2225 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 9920 | Gm9920 | ENSMUSG00000053749 | -143.5kb | Anti-sense | Chr. 15 - 55099917 - 55113582 on NCBI Build GRCm38 |
| Downstream |
collagen, type XIV, alpha 1 |
Col14a1 |
5730412L22Rik, AW108078 |
48.4kb | Sense | Chr. 15 - 55307706 - 55520803 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Deptor
Genomic Map