Assay Details
| Gene Symbol: 4833406L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014657.1 | - | 1 | 649 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11820 | Gm11820 | OTTMUSG00000004463 | -737.3kb | Sense | Chr. 4 - 13076140 - 13117434 on NCBI Build GRCm38 |
| Downstream | predicted gene 11823 | Gm11823 |
D930021N14, OTTMUSG00000004480 |
64.2kb | Anti-sense | Chr. 4 - 13919954 - 13995193 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4833406L22Rik
Genomic Map