Assay Details
| Gene Symbol: 4921504P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014816.1 | - | 1 | 447 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 8b | Zbtb8b |
AV220464, BC023839, BOZF1, Zbtb8 |
-13.2kb | Sense | Chr. 4 - 129425765 - 129446972 on NCBI Build GRCm38 |
| Downstream | BSD domain containing 1 | Bsdc1 |
1110063F24Rik, AW011758 |
155.0 | Anti-sense | Chr. 4 - 129461679 - 129488498 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4921504P20Rik
Genomic Map