Assay Details
| Gene Symbol: 4930405G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015097.1 | - | 1 | 744 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 16157 | Gm16157 | OTTMUSG00000031023 | -39.4kb | Sense | Chr. 7 - 68266339 - 68276594 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933436H12 gene | 4933436H12Rik | 100.4kb | Anti-sense | Chr. 7 - 68417424 - 68426133 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930405G09Rik
Genomic Map