Assay Details
| Gene Symbol: 4930423D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015187.1 | - | 1 | 1546 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase 9 | Map3k9 |
E130314H24Rik, Mlk1, Prke1 |
Anti-sense | Chr. 12 - 81714950 - 81782549 on NCBI Build GRCm38 | |
| Downstream | pecanex homolog (Drosophila) | Pcnx |
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1 |
77.0kb | Sense | Chr. 12 - 81860028 - 82000924 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930423D22Rik
Genomic Map