Assay Details
| Gene Symbol: P2rx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF310647.1 | - | 1 | 36 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, Ca++ transporting, ubiquitous |
Atp2a3 |
SERCA3b, Serca3 |
-6.0kb | Sense | Chr. 11 - 72961169 - 72993043 on NCBI Build GRCm38 |
| Downstream |
calcium/calmodulin-dependent protein kinase kinase 1, alpha |
Camkk1 |
AI846603, CaMKKalpha, Camkk |
16.2kb | Sense | Chr. 11 - 73015306 - 73042073 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

P2rx1
Genomic Map