Assay Details
| Gene Symbol: Gml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177524.1 | NP_001170995.1 | 2 - 3 | 341 | 66 | |
| GenBank mRNA | AJ242831.1 | - | 1 - 2 | 400 | 66 | |
| CJ042352.1 | - | 2 - 3 | 66 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
lymphocyte antigen 6 complex, locus K |
Ly6k |
2410015A16Rik, 3110035B01Rik, mLy-6K |
-16.4kb | Sense | Chr. 15 - 74796874 - 74799968 on NCBI Build GRCm38 |
| Downstream | hematopoietic cell transcript 1 | Hemt1 |
1700057K19Rik, HemT |
1.4kb | Anti-sense | Chr. 15 - 74818880 - 74846164 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gml
Genomic Map