Assay Details
| Gene Symbol: Socs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005238.1 | - | 1 | 346 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cerebellin 2 precursor protein | Cbln2 |
6330593N19Rik, A730004O05 |
-2.1mb | Anti-sense | Chr. 18 - 86711053 - 86718285 on NCBI Build GRCm38 |
| Downstream | rotatin | Rttn |
4921538A15Rik, AI666264, C530033I08Rik, D230040K24 |
105.9kb | Anti-sense | Chr. 18 - 88971738 - 89131016 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Socs6
Genomic Map