Assay Details
| Gene Symbol: 2610318M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019187.1 | - | 1 | 47 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102632156 | LOC102632156 | Sense | Chr. 14 - 8367437 - 8371150 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 4930452B06 gene | 4930452B06Rik |
4933406L09Rik, AI854000 |
62.6kb | Anti-sense | Chr. 14 - 8431171 - 8666502 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610318M16Rik
Genomic Map