Assay Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019287.1 | - | 1 | 50 | 60 | |
| AK028267.1 | - | 1 | 98 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
LSM6 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Lsm6 |
1500031N17Rik, 2410088K19Rik, AI747288 |
-207.1kb | Anti-sense | Chr. 8 - 78804868 - 78821152 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700011L22 gene | 1700011L22Rik | 182.0kb | Anti-sense | Chr. 8 - 79210423 - 79248897 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp827
Genomic Map