Assay Details
| Gene Symbol: 4930552P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019761.1 | - | 1 | 346 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GLIS family zinc finger 1 | Glis1 |
Gli5, Gli6, GliH1 |
Anti-sense | Chr. 4 - 107426640 - 107635061 on NCBI Build GRCm38 | |
| Downstream |
DMRT-like family B with proline-rich C-terminal, 1 |
Dmrtb1 |
Dmrt6, Prp13 |
79.1kb | Sense | Chr. 4 - 107676288 - 107684243 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930552P06Rik
Genomic Map