Assay Details
| Gene Symbol: 5430409L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019945.1 | - | 1 | 144 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 365-2 | Mir365-2 |
Mirn365-2, mir-365-2 |
-233.6kb | Anti-sense | Chr. 11 - 79726400 - 79726511 on NCBI Build GRCm38 |
| Downstream | suppressor of zeste 12 homolog (Drosophila) | Suz12 |
2610028O16Rik, AI195385, AU016842, AW536442, D11Ertd530e, mKIAA0160 |
32.6kb | Anti-sense | Chr. 11 - 79993106 - 80034123 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430409L15Rik
Genomic Map