Assay Details
| Gene Symbol: 6030442H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020062.1 | - | 1 | 1049 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13575 | Gm13575 | OTTMUSG00000012829 | -623.4kb | Sense | Chr. 2 - 64363699 - 64393881 on NCBI Build GRCm38 |
| Downstream | Cobl-like 1 | Cobll1 |
1810047P18Rik, Coblr1, D430044D16Rik |
69.8kb | Sense | Chr. 2 - 65088339 - 65239480 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6030442H21Rik
Genomic Map