Assay Details
| Gene Symbol: 6030492E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020086.1 | - | 1 | 849 | 108 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DV073193.1 | - | 393 | Chr. - - [-] on Build | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box P1 | Foxp1 |
3110052D19Rik, 4932443N09Rik, AI461938, AW494214 |
Sense | Chr. 6 - 98925342 - 99523024 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 19665 |
Gm19665 | 99.1kb | Sense | Chr. 6 - 99097669 - 99108316 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6030492E11Rik
Genomic Map