Assay Details
| Gene Symbol: 9230106L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020322.1 | - | 1 | 849 | 72 | |
| AV380998.1 | - | 1 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transformation related protein 63 | Trp63 |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
-10.3kb | Anti-sense | Chr. 16 - 25683763 - 25892103 on NCBI Build GRCm38 |
| Downstream | predicted gene 4524 | Gm4524 | 47.0kb | Anti-sense | Chr. 16 - 25950435 - 25960356 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9230106L01Rik
Genomic Map