Assay Details
| Gene Symbol: 6530411M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020377.1 | - | 1 | 856 | 68 | |
| AK035915.1 | - | 1 | 757 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700010I14 gene | 1700010I14Rik | -158.8kb | Anti-sense | Chr. 17 - 8988320 - 9008319 on NCBI Build GRCm38 | |
| Downstream |
poly(A) binding protein, cytoplasmic 6 |
Pabpc6 |
4932702K14Rik, AI428050, Pabpc3 |
498.5kb | Sense | Chr. 17 - 9666497 - 9669704 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6530411M01Rik
Genomic Map