Assay Details
| Gene Symbol: 9430027B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020441.1 | - | 1 | 747 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 30 (zinc transporter), member 9 |
Slc30a9 |
2310024J23Rik, AL024256, GAC63, HUEL |
-24.3kb | Anti-sense | Chr. 5 - 67306937 - 67356834 on NCBI Build GRCm38 |
| Downstream | BEN domain containing 4 | Bend4 |
D330027G24Rik, EG666938 |
10.0kb | Sense | Chr. 5 - 67392147 - 67428250 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430027B09Rik
Genomic Map