Assay Details
| Gene Symbol: 9430085L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020505.1 | - | 1 | 746 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6878 | Gm6878 | EG628416 | -26.1kb | Anti-sense | Chr. 14 - 67304888 - 67314711 on NCBI Build GRCm38 |
| Downstream | predicted gene 10860 | Gm10860 | ENSMUSG00000075548 | 88.3kb | Anti-sense | Chr. 14 - 67430194 - 67437171 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430085L16Rik
Genomic Map