Assay Details
| Gene Symbol: 9530062E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020618.1 | - | 1 | 252 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | diaphanous homolog 2 (Drosophila) | Diap2 |
Dia3, Diaph2, Drf2, E430022I22Rik |
Sense | Chr. X - 129749668 - 130465834 on NCBI Build GRCm38 | |
| Downstream | protocadherin 19 | Pcdh19 |
B530002L05Rik, Gm717, mKIAA1313 |
3.2mb | Anti-sense | Chr. X - 133582860 - 133688993 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530062E16Rik
Genomic Map