Assay Details
| Gene Symbol: 9530086O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020657.1 | - | 1 | 744 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26911 |
Gm26911 | -93.8kb | Anti-sense | Chr. 6 - 101059133 - 101221620 on NCBI Build GRCm38 | |
| Downstream | predicted gene 9871 | Gm9871 |
0610005I04, EG207157, ENSMUSG00000052236 |
457.7kb | Anti-sense | Chr. 6 - 101774249 - 101801982 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530086O07Rik
Genomic Map