Assay Details
| Gene Symbol: A930003G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020804.1 | - | 1 | 152 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
vomeronasal 2, receptor 56 |
Vmn2r56 | EG629079 | -70.2kb | Anti-sense | Chr. 7 - 12693860 - 12733320 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 110 | Zfp110 |
2900024E01Rik, N28112, NRIF, Nrif1 |
30.2kb | Sense | Chr. 7 - 12834810 - 12850584 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930003G23Rik
Genomic Map