Assay Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020822.1 | - | 1 | 546 | 185 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 162, member A |
Fam162a |
2310056P07Rik, HGTD-P |
-565.0 | Anti-sense | Chr. 16 - 36043844 - 36071515 on NCBI Build GRCm38 |
| Downstream | cystatin A1 | Csta1 | Csta | 47.2kb | Anti-sense | Chr. 16 - 36119946 - 36131189 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc58
Genomic Map