Assay Details
| Gene Symbol: 3110035F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014124.1 | - | 1 | 249 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glypican 6 | Gpc6 | Sense | Chr. 14 - 116921001 - 117979529 on NCBI Build GRCm38 | ||
| Downstream | microRNA 6239 | Mir6239 |
Gm28025, mmu-mir-6239 |
287.7kb | Anti-sense | Chr. 14 - 117953743 - 117953847 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3110035F07Rik
Genomic Map