Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | S100 calcium binding protein A4 | S100a4 |
18A2, 42a, Capl, FSp1, Mts1, PeL98, metastasin, pk9a |
Sense | Chr. 3 - 90603770 - 90606045 on NCBI Build GRCm38 | |
| Downstream | S100 calcium binding protein A5 | S100a5 | S100D9 | 2.5kb | Sense | Chr. 3 - 90606479 - 90611784 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map