Assay Details
| Gene Symbol: Gm16702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045795.1 | 2 - 3 | 696 | 68 | ||
| GenBank mRNA | AK028328.1 | - | 2 - 3 | 697 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | brachyury 2 | T2 | brachyury2 | Anti-sense | Chr. 17 - 8372015 - 8422728 on NCBI Build GRCm38 | |
| Downstream | brachyury | T |
Bra, D17Mit170, Low, Lr, T1, Tl2, Tl3, cou, me75 |
36.0kb | Anti-sense | Chr. 17 - 8422178 - 8442496 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16702
Genomic Map