Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10677 | Gm10677 | ENSMUSG00000074363 | -17.7kb | Anti-sense | Chr. 9 - 47799160 - 47818208 on NCBI Build GRCm38 |
| Downstream |
neurexophilin and PC-esterase domain family, member 4 |
Nxpe4 |
C130036J11, D930028F11Rik, Fam55d |
325.5kb | Sense | Chr. 9 - 48162001 - 48400025 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map