Assay Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022735.1 | - | 1 | 451 | 103 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BG297260.1 | - | 450 | Chr. - - [-] on Build | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dipeptidylpeptidase 3 | Dpp3 |
4930533O14Rik, C86324 |
-1.4kb | Sense | Chr. 19 - 4907229 - 4928287 on NCBI Build GRCm38 |
| Downstream | mitochondrial ribosomal protein L11 | Mrpl11 |
2410001P07Rik, L11mt |
31.1kb | Anti-sense | Chr. 19 - 4962152 - 4966995 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Peli3
Genomic Map