Assay Details
| Gene Symbol: Gm14204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040358.1 | 3 | 4381 | 69 | ||
| GenBank mRNA | AK034893.1 | - | 1 | 1346 | 69 | |
| BC072662.1 | - | 3 | 4381 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 40 | Arhgap40 |
C20orf95, Gm14203, OTTMUSG00000015933 |
-47.5kb | Anti-sense | Chr. 2 - 158512796 - 158550628 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 32 (GABA vesicular transporter), member 1 |
Slc32a1 |
R75019, VGAT, Viaat |
10.2kb | Anti-sense | Chr. 2 - 158610758 - 158615747 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm14204
Genomic Map