Assay Details
| Gene Symbol: Gm32486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035251.1 | - | 1 - 2 | 96 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PWWP domain containing 2B | Pwwp2b |
AI594893, D7Ertd517e, D930023J19Rik, Pwwp2 |
-25.6kb | Anti-sense | Chr. 7 - 139248482 - 139267253 on NCBI Build GRCm38 |
| Downstream | inositol polyphosphate-5-phosphatase A | Inpp5a | 92.2kb | Anti-sense | Chr. 7 - 139389109 - 139579652 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32486
Genomic Map