Assay Details
| Gene Symbol: Gm16206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035416.1 | - | 1437 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NHL repeat containing 3 | Nhlrc3 |
4833441N19, 8030451K01Rik, mKIAA4083 |
Anti-sense | Chr. 3 - 53451996 - 53463258 on NCBI Build GRCm38 | |
| Downstream | proline and serine rich 1 | Proser1 |
2810046L04Rik, 9330161F11 |
6.0kb | Sense | Chr. 3 - 53463817 - 53481755 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16206
Genomic Map