Assay Details
| Gene Symbol: Gm13415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037056.1 | - | 2 - 3 | 283 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 8 | Pax8 | Pax-8 | Anti-sense | Chr. 2 - 24420551 - 24480092 on NCBI Build GRCm38 | |
| Downstream |
calcium channel, voltage-dependent, N type, alpha 1B subunit |
Cacna1b |
AW050276, AW060892, AW822256, Cav2.2, Cchn1a, alpha(1B) |
142.6kb | Anti-sense | Chr. 2 - 24603860 - 24763195 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13415
Genomic Map