Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | notch 2 | Notch2 |
AI853703, N2 |
Sense | Chr. 3 - 98013538 - 98150367 on NCBI Build GRCm38 | |
| Downstream | a disintegrin and metallopeptidase domain 30 | Adam30 |
4933424D07Rik, svph4 |
36.1kb | Sense | Chr. 3 - 98160812 - 98163173 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map