Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15689 | Gm15689 | OTTMUSG00000025312 | -36.7kb | Anti-sense | Chr. 3 - 141643655 - 141683463 on NCBI Build GRCm38 |
| Downstream |
bone morphogenetic protein receptor, type 1B |
Bmpr1b |
AI385617, ALK-6, AV355320, Acvrlk6, Alk6, BMPR-1B, BMPR-IB, CFK-43a, SKR6 |
116.3kb | Anti-sense | Chr. 3 - 141837136 - 142169590 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map