| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200043153.mm.1 |
NR_040358.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043153.mm.1 |
NR_040358.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043153.mm.1 |
NR_040359.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043153.mm.1 |
NR_040359.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043153.mm.1 |
TC0200005041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043142.mm.1 |
NR_040359.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043142.mm.1 |
NR_040359.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043142.mm.1 |
TC0200005041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043154.mm.1 |
NR_040358.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043154.mm.1 |
NR_040358.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043154.mm.1 |
NR_040359.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043154.mm.1 |
NR_040359.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043154.mm.1 |
TC0200005041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043140.mm.1 |
NR_040359.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043140.mm.1 |
NR_040359.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043140.mm.1 |
TC0200005041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043151.mm.1 |
NR_040358.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043151.mm.1 |
NR_040358.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043151.mm.1 |
NR_040359.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043151.mm.1 |
NR_040359.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043151.mm.1 |
TC0200005041.mm.1 |
MTA TC |