Assay Details
| Gene Symbol: Gm32496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143350.1 | - | 1 - 2 | 130 | 160 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stathmin-like 2 | Stmn2 |
AI159727, SCG10, Scgn10, Stmb2 |
-66.4kb | Sense | Chr. 3 - 8509527 - 8561604 on NCBI Build GRCm38 |
| Downstream | hairy/enhancer-of-split related with YRPW motif 1 | Hey1 |
AI316788, AI414254, CHF2, HRT1, Herp2, Hesr1, bHLHb31, hesr-1 |
34.9kb | Anti-sense | Chr. 3 - 8663359 - 8667038 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm32496
Genomic Map