Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin alpha 7-gamma | Pcdha7-g | Anti-sense | Chr. 18 - 36973802 - 37841873 on NCBI Build GRCm38 | ||
| Downstream | protocadherin beta 1 | Pcdhb1 | PcdhbA | 112.3kb | Anti-sense | Chr. 18 - 37264998 - 37267454 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map