| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500012134.mm.1 |
NM_001168274.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012134.mm.1 |
NM_001168274.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012134.mm.1 |
TC1500002344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012130.mm.1 |
NM_001168274.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012130.mm.1 |
NM_001168274.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012130.mm.1 |
NM_177912.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012130.mm.1 |
NM_177912.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012130.mm.1 |
TC1500002344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012133.mm.1 |
NM_001168274.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012133.mm.1 |
NM_001168274.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012133.mm.1 |
TC1500002344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012131.mm.1 |
NM_001168274.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012131.mm.1 |
NM_001168274.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012131.mm.1 |
NM_177912.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012131.mm.1 |
NM_177912.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012131.mm.1 |
TC1500002344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500012135.mm.1 |
NM_001168274.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012135.mm.1 |
NM_001168274.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012135.mm.1 |
NM_177912.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012135.mm.1 |
NM_177912.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500012135.mm.1 |
TC1500002344.mm.1 |
MTA TC |