Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102632465 | LOC102632465 | -386.9kb | Anti-sense | Chr. 2 - 6466079 - 6478619 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 5031426D15 gene | 5031426D15Rik | 56.6kb | Sense | Chr. 2 - 6922702 - 6928722 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map