Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
neurotrophic tyrosine kinase, receptor, type 2 |
Ntrk2 |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Sense | Chr. 13 - 58806569 - 59133970 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 34245 |
Gm34245 | 203.4kb | Anti-sense | Chr. 13 - 59070788 - 59082920 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map