Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | roundabout homolog 2 (Drosophila) | Robo2 |
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568 |
Sense | Chr. 16 - 73892061 - 75447298 on NCBI Build GRCm38 | |
| Downstream | microRNA 691 | Mir691 |
Mirn691, mir-691, mmu-mir-691 |
164.4kb | Sense | Chr. 16 - 74341990 - 74342067 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map