Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear receptor coactivator 1 | Ncoa1 |
KAT13A, SRC-1, SRC1, bHLHe74 |
Sense | Chr. 12 - 4247362 - 4477210 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31938 |
Gm31938 | 140.2kb | Anti-sense | Chr. 12 - 4523853 - 4529122 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map