Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15506 | Gm15506 | OTTMUSG00000023003 | Sense | Chr. 7 - 98682331 - 98703391 on NCBI Build GRCm38 | |
| Downstream |
protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor) |
Prkrir |
2900052B10Rik, Dap4, Rpkrir |
3.3kb | Anti-sense | Chr. 7 - 98703230 - 98718062 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map