Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 301 | Mir301 |
Mirn301, mir-301a, mmu-mir-301 |
Sense | Chr. 11 - 87113004 - 87113089 on NCBI Build GRCm38 | |
| Downstream | tripartite motif-containing 37 | Trim37 |
1110032A10Rik, 2810004E07Rik, AI848587, AU043018, MUL, TEF3, mKIAA0898 |
13.4kb | Sense | Chr. 11 - 87127051 - 87250874 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map