Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 196b | Mir196b |
Mirn196b, mir-196b |
-8.8kb | Sense | Chr. 6 - 52230081 - 52230165 on NCBI Build GRCm38 |
| Downstream | homeobox A11 | Hoxa11 |
Hox-1.9, Hoxa-11 |
2.5kb | Sense | Chr. 6 - 52242105 - 52245810 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map