Assay Details
| Gene Symbol: Gm16861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037666.1 | 2 - 3 | 514 | 57 | ||
| GenBank mRNA | AK078615.2 | - | 2 - 3 | 516 | 57 | |
| BC061511.1 | - | 2 - 3 | 562 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mahogunin, ring finger 1 |
Mgrn1 |
2610042J20Rik, mKIAA0544, md, nc |
Anti-sense | Chr. 16 - 4886100 - 4938296 on NCBI Build GRCm38 | |
| Downstream | ankyrin repeat and sterile alpha motif domain containing 3 | Anks3 |
2700067D09Rik, C81345, mKIAA1977 |
2.3kb | Sense | Chr. 16 - 4941415 - 4964327 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16861
Genomic Map