Assay Details
| Gene Symbol: Gm11820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139819.1 | - | 5 - 6 | 762 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 42256 |
Gm42256 | -75.3kb | Sense | Chr. 4 - 12981683 - 13025788 on NCBI Build GRCm38 | |
| Downstream |
runt-related transcription factor 1; translocated to, 1 (cyclin D-related) |
Runx1t1 |
Cbfa2t1h, ETO, MTG8 |
641.3kb | Sense | Chr. 4 - 13743182 - 13895056 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11820
Genomic Map