Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | B cell CLL/lymphoma 11A (zinc finger protein) | Bcl11a |
2810047E18Rik, BCL-11A, Ctip1, D930021L15Rik, Evi9, Evi9a, Evi9b, Evi9c, mKIAA1809 |
-6.6kb | Sense | Chr. 11 - 24076564 - 24173558 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933430M04 gene | 4933430M04Rik | 301.1kb | Sense | Chr. 11 - 24481909 - 24569951 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map