Assay Details
| Gene Symbol: Gm12069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158601.1 | - | 2 - 3 | 171 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10466 | Gm10466 | ENSMUSG00000073103 | -590.7kb | Anti-sense | Chr. 11 - 24721006 - 24730722 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933427E13 gene | 4933427E13Rik | 3.3kb | Sense | Chr. 11 - 25326107 - 25367137 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12069
Genomic Map