Assay Details
| Gene Symbol: Gm10814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045783.1 | 1 - 2 | 59 | 79 | ||
| GenBank mRNA | AK160921.1 | - | 1 - 2 | 59 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calpain 1 | Capn1 |
Capa-1, Capa1, mu-calpin |
Anti-sense | Chr. 19 - 5988545 - 6015825 on NCBI Build GRCm38 | |
| Downstream |
synovial apoptosis inhibitor 1, synoviolin |
Syvn1 |
1200010C09Rik, AW211966, C85322, D530017H19Rik, Hrd1 |
30.7kb | Sense | Chr. 19 - 6045030 - 6053718 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10814
Genomic Map