Assay Details
| Gene Symbol: Gm33100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171414.1 | - | 2 - 3 | 207 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | annexin A3 | Anxa3 | Anx3 | -33.7kb | Sense | Chr. 5 - 96793385 - 96845968 on NCBI Build GRCm38 |
| Downstream | predicted gene 8013 | Gm8013 | EG666266 | 26.1kb | Sense | Chr. 5 - 96911764 - 96934867 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm33100
Genomic Map